missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-amino-3-pyridyl)acrylate

$287.85 - $1,266.82

Chemical Identifiers

CAS 104830-07-1
Molecular Formula C10H12N2O2
Formula Weight 192.22
MDL Number MFCD07781149
Synonym 3-(4-Amino-3-pyridyl)acrylic acid ethyl ester
Chemical Name or Material Ethyl 3-(4-amino-3-pyridyl)acrylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5003706 SDS Alfa Aesar™
H5003706
5g Each for $1,266.82
AAH5003703 SDS Alfa Aesar™
H5003703
1g Each for $287.85
Specifications Description & Specifications

Chemical Identifiers

104830-07-1
192.22
3-(4-Amino-3-pyridyl)acrylic acid ethyl ester
C10H12N2O2
MFCD07781149
Ethyl 3-(4-amino-3-pyridyl)acrylate

Specifications

104830-07-1
192.22
3-(4-Amino-3-pyridyl)acrylic acid ethyl ester
Ethyl 3-(4-amino-3-pyridyl)acrylate
C10H12N2O2
MFCD07781149
5g