missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,3-diethoxypropionate, 96%

Manufacturer:  Alfa Aesar™ L0956218

 View more versions of this product

Catalog No. AAL0956218



Specifications

Specifications

Ethyl 3,3-diethoxypropionate
92°C to 93°C (13mmHg)
10601-80-6
MFCD00009865
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
CCOC(CC(=O)OCC)OCC
190.239
190.24
93°C (199°F)
50g
C9H18O4
1772110
SIALOQYKFQEKOG-UHFFFAOYSA-N
ethyl 3,3-diethoxypropanoate
66389
96%

Chemical Identifiers

10601-80-6
190.239
SIALOQYKFQEKOG-UHFFFAOYSA-N
66389
CCOC(CC(=O)OCC)OCC
C9H18O4
MFCD00009865
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
ethyl 3,3-diethoxypropanoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C

Documents