Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-[3-(aminomethyl)phenyl]-2-methylpropionate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 3-[3-(aminomethyl)phenyl]-2-methylpropionate
Molecular Formula C13H19NO2
Formula Weight 221.3
MDL Number MFCD08064280
Synonym 3-[3-(Aminomethyl)phenyl]-2-methylpropionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5100003

  SDS
alfa aesar™
H5100003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51000MD

  SDS
alfa aesar™
H51000MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C13H19NO2
MDL Number MFCD08064280
Assay 95%
Chemical Name or Material Ethyl 3-[3-(aminomethyl)phenyl]-2-methylpropionate
Health Hazard 1 Warning
Quantity 1g
Synonym 3-[3-(Aminomethyl)phenyl]-2-methylpropionic acid ethyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.