Please login to your online account to display your discounted pricing

Ethyl 3-[3,5-di(trifluoromethyl)phenyl]-3-oxopropanoate, 90%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate
CAS 175278-02-1
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C13H10F6O3
Linear Formula (CF3)2C6H3C(O)CH2CO2CH2CH3
Formula Weight 328.21
MDL Number MFCD00052321
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00364DA

  SDS
fisher scientific company
TL00364DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TL00364EA

  SDS
fisher scientific company
TL00364EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 90%
Formula Weight 328.21
Quantity 1g
CAS Min % 90.0
Chemical Name or Material Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate
Packaging Amber Glass Bottle
Assay Percent Range 90%
Molecular Formula C13H10F6O3
Linear Formula (CF3)2C6H3C(O)CH2CO2CH2CH3
MDL Number MFCD00052321
CAS 175278-02-1