Please login to your online account to display your discounted pricing

Ethyl 3-[3,5-di(trifluoromethyl)phenyl]-3-oxopropanoate, 90%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  TL00364EA

 View more versions of this product

Catalog No. TL00364EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 90%
Formula Weight 328.21
Quantity 10g
CAS Min % 90.0
Assay Percent Range 90%
Molecular Formula C13H10F6O3
Chemical Name or Material Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate
MDL Number MFCD00052321
Linear Formula (CF3)2C6H3C(O)CH2CO2CH2CH3
CAS 175278-02-1

Specifications

Chemical Name or Material Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate
CAS 175278-02-1
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C13H10F6O3
Linear Formula (CF3)2C6H3C(O)CH2CO2CH2CH3
Formula Weight 328.21
MDL Number MFCD00052321