Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(3,4,5-trimethoxyphenyl)propionate, 98%

$72.18 - $283.27

Chemical Identifiers

Chemical Name or Material Ethyl 3- (3,4,5-trimethoxyphenyl)propionate
CAS 70311-20-5
Molecular Formula C14H20O5
Formula Weight 268.31
MDL Number MFCD00017522
Synonym 3-(3,4,5-Trimethoxyphenyl)propionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1245906

  SDS
alfa aesar™
A1245906
5g Each for $72.18

AAA1245914

  SDS
alfa aesar™
A1245914
25g Each for $283.27
Description & Specifications

Specifications

Formula Weight 268.31
Synonym 3-(3,4,5-Trimethoxyphenyl)propionic acid ethyl ester
Chemical Name or Material Ethyl 3- (3,4,5-trimethoxyphenyl)propionate
Quantity 5g
CAS 70311-20-5
Assay 98%
Molecular Formula C14H20O5
MDL Number MFCD00017522