missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-trimethylacetamido-3-pyridyl)acrylate

$240.84 - $1,066.59

Chemical Identifiers

CAS 882029-13-2
Molecular Formula C15H20N2O3
Formula Weight 276.34
MDL Number MFCD07781145
Synonym Ethyl 3-(2-pivaloylamino-3-pyridyl)acrylate; 3-(2-Trimethylacetamido-3-pyridyl)acrylic acid ethyl ester
Chemical Name or Material Ethyl 3-(2-trimethylacetamido-3-pyridyl)acrylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5001706 SDS Alfa Aesar™
H5001706
5g Each for $1,066.59
AAH5001703 SDS Alfa Aesar™
H5001703
1g Each for $240.84
Specifications Description & Specifications

Chemical Identifiers

882029-13-2
276.34
Ethyl 3-(2-pivaloylamino-3-pyridyl)acrylate; 3-(2-Trimethylacetamido-3-pyridyl)acrylic acid ethyl ester
C15H20N2O3
MFCD07781145
Ethyl 3-(2-trimethylacetamido-3-pyridyl)acrylate

Specifications

882029-13-2
276.34
Ethyl 3-(2-pivaloylamino-3-pyridyl)acrylate; 3-(2-Trimethylacetamido-3-pyridyl)acrylic acid ethyl ester
Ethyl 3-(2-trimethylacetamido-3-pyridyl)acrylate
C15H20N2O3
MFCD07781145
5g