Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-thienyl)piperazine-1-carboxylate, 95%

$500.24 - $500.24

Chemical Identifiers

Chemical Name or Material Ethyl 3-(2-thienyl)piperazine-1-carboxylate
CAS 85803-50-5
Molecular Formula C11H16N2O2S
Formula Weight 240.33
MDL Number MFCD06245431
Synonym 1-Ethoxycarbonyl-3-(2-thienyl)piperazine; 3-(2-Thienyl)piperazine-1-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26527MD

  SDS
alfa aesar™
H26527MD
250mg Each for $500.24
Description & Specifications

Specifications

Formula Weight 240.33
MDL Number MFCD06245431
Molecular Formula C11H16N2O2S
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 250mg
Assay 95%
CAS 85803-50-5
Synonym 1-Ethoxycarbonyl-3-(2-thienyl)piperazine; 3-(2-Thienyl)piperazine-1-carboxylic acid ethyl ester
Chemical Name or Material Ethyl 3-(2-thienyl)piperazine-1-carboxylate
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.