missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-methoxy-3-pyridyl)acrylate

$266.30 - $1,183.70

Chemical Identifiers

CAS 912760-91-9
Molecular Formula C11H13NO3
Formula Weight 207.23
MDL Number MFCD07781176
Synonym 3-(2-Methoxy-3-pyridyl)acrylic acid ethyl ester
Chemical Name or Material Ethyl 3-(2-methoxy-3-pyridyl)acrylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5011903 SDS Alfa Aesar™
H5011903
1g Each for $266.30
AAH5011906 SDS Alfa Aesar™
H5011906
5g Each for $1,183.70
Specifications Description & Specifications

Chemical Identifiers

912760-91-9
207.23
3-(2-Methoxy-3-pyridyl)acrylic acid ethyl ester
C11H13NO3
MFCD07781176
Ethyl 3-(2-methoxy-3-pyridyl)acrylate

Specifications

912760-91-9
207.23
3-(2-Methoxy-3-pyridyl)acrylic acid ethyl ester
Ethyl 3-(2-methoxy-3-pyridyl)acrylate
C11H13NO3
MFCD07781176
1g