Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-chloro-3-pyridyl)acrylate

$275.53 - $1,219.11

Chemical Identifiers

Chemical Name or Material Ethyl 3-(2-chloro-3-pyridyl)acrylate
CAS 104830-08-2
Molecular Formula C10H10ClNO2
Formula Weight 211.65
MDL Number MFCD05663505
Synonym 3-(2-Chloro-3-pyridyl)acrylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5005003

 
alfa aesar™
H5005003
1g Each for $275.53

AAH5005006

 
alfa aesar™
H5005006
5g Each for $1,219.11
Description & Specifications

Specifications

Formula Weight 211.65
Chemical Name or Material Ethyl 3-(2-chloro-3-pyridyl)acrylate
Molecular Formula C10H10ClNO2
MDL Number MFCD05663505
CAS 104830-08-2
Synonym 3-(2-Chloro-3-pyridyl)acrylic acid ethyl ester
Quantity 1g