missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-bromophenyl)propionate, 98%

Manufacturer:  Alfa Aesar™ H6350503

 View more versions of this product

Catalog No. AAH6350503



Specifications

Specifications

Ethyl 3-(2-bromophenyl)propionate
1g
C11H13BrO2
ethyl 3-2-bromophenyl propanoate, 3-2-bromo-phenyl-propionic acid ethyl ester, 3-2-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 2-bromo-, ethyl ester, ksc497c5h, ethyl3-2-bromophenyl propanoate, ethyl 2-bromo-3-benzenepropanoate, 2-bromo-hydrocinnamic acid ethyl ester, 3-2-bromophenyl propanoic acid ethyl ester
CCOC(=O)CCC1=CC=CC=C1Br
257.127
257.13
98%
Yellow
135613-33-1
MFCD07772962
ZKICRDRBPPOQLM-UHFFFAOYSA-N
ethyl 3-(2-bromophenyl)propanoate
10611137
Liquid

Chemical Identifiers

135613-33-1
257.127
ZKICRDRBPPOQLM-UHFFFAOYSA-N
10611137
CCOC(=O)CCC1=CC=CC=C1Br
C11H13BrO2
MFCD07772962
ethyl 3-2-bromophenyl propanoate, 3-2-bromo-phenyl-propionic acid ethyl ester, 3-2-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 2-bromo-, ethyl ester, ksc497c5h, ethyl3-2-bromophenyl propanoate, ethyl 2-bromo-3-benzenepropanoate, 2-bromo-hydrocinnamic acid ethyl ester, 3-2-bromophenyl propanoic acid ethyl ester
ethyl 3-(2-bromophenyl)propanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents