Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(2-amino-3-pyridyl)acrylate, 95%

$1,226.22 - $1,226.22

Chemical Identifiers

Chemical Name or Material Ethyl 3-(2-amino-3-pyridyl)acrylate
CAS 104830-01-5
Molecular Formula C10H12N2O2
Formula Weight 192.22g/mol
MDL Number MFCD07781146
Synonym 3-(2-Amino-3-pyridyl)acrylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5001806

 
alfa aesar™
H50018-06
Each for $1,226.22
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Ethyl 3-(2-amino-3-pyridyl)acrylate
CAS 104830-01-5
Assay 95%
Molecular Formula C10H12N2O2
Formula Weight 192.22g/mol
MDL Number MFCD07781146
Synonym 3-(2-Amino-3-pyridyl)acrylic acid ethyl ester