Please login to your online account to display your discounted pricing

Ethyl 3-[2,6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  335410050

 View more versions of this product

Catalog No. AC335410050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 280.08
Quantity 5g
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Ethyl 2, 6-dichloro-5-fluornicotinoylacetate
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS 96568-04-6
Name Note 97%
Chemical Name or Material Ethyl 3-[2, 6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate
MDL Number MFCD00799535
Assay Percent Range 97%
Molecular Formula C10H8Cl2FNO3

Specifications

Chemical Name or Material Ethyl 3-[2, 6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate
Name Note 97%
CAS 96568-04-6
Assay Percent Range 97%
Molecular Formula C10H8Cl2FNO3
Formula Weight 280.08
MDL Number MFCD00799535
Synonym Ethyl 2, 6-dichloro-5-fluornicotinoylacetate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents