Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(trifluoromethyl)benzoate, 98%

$124.68 - $1,847.14

Chemical Identifiers

Chemical Name or Material Ethyl 2-(trifluoromethyl)benzoate
CAS 577-62-8
Molecular Formula C10H9F3O2
Formula Weight 218.18
MDL Number MFCD00013556
Beilstein 3290053
Synonym 2-(Trifluoromethyl)benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1621506

 
alfa aesar™
A1621506
5g Each for $124.68

AAA1621514

 
alfa aesar™
A1621514
25g Each for $563.38

AAA1621522

 
alfa aesar™
A1621522
100g Each for $1,847.14
Description & Specifications

Specifications

Formula Weight 218.18
Chemical Name or Material Ethyl 2-(trifluoromethyl)benzoate
Assay 98%
Molecular Formula C10H9F3O2
MDL Number MFCD00013556
CAS 577-62-8
Beilstein 3290053
Synonym 2-(Trifluoromethyl)benzoic acid ethyl ester
Boiling Point 90° to 91°C (10mmHg)
Quantity 5g