missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3206503

 View more versions of this product

Catalog No. AAH3206503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate
54°C (0.5 mmHg)
MFCD07367386
ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester
CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F
234.178
234.18
306960-67-8
C9H9F3N2O2
1 g
LYEKBQSEKSEXLT-UHFFFAOYSA-N
ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
10609709
97%

Chemical Identifiers

306960-67-8
234.178
LYEKBQSEKSEXLT-UHFFFAOYSA-N
10609709
CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F
C9H9F3N2O2
MFCD07367386
ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester
ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only