Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-thiopheneacetate, 97%

$58.49 - $977.45

Chemical Identifiers

Chemical Name or Material Ethyl 2-thiopheneacetate
CAS 57382-97-5
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00005457
Beilstein 127918
Synonym 2-Thiopheneacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1589609

 
alfa aesar™
A1589609
10g Each for $58.49

AAA1589618

 
alfa aesar™
A1589618
50g Each for $227.81

AAA1589630

 
alfa aesar™
A1589630
250g Each for $977.45
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material Ethyl 2-thiopheneacetate
Assay 97%
Molecular Formula C8H10O2S
MDL Number MFCD00005457
CAS 57382-97-5
Beilstein 127918
Synonym 2-Thiopheneacetic acid ethyl ester
Flash Point 104°C (219°F)
Boiling Point 109° to 111°C (11mmHg)
Quantity 10g