Please login to your online account to display your discounted pricing

Ethyl 2-pyridylacetate 98%, ACROS Organics™

$81.29 - $81.29

Chemical Identifiers

Chemical Name or Material Ethyl 2-pyridylacetate
Name Note 98%
CAS 2739-98-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H11NO2
Linear Formula (C5H4N)CH2COOC2H5
Formula Weight 165.19
MDL Number MFCD00006359
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118910050

  SDS
fisher scientific acros
118910050
Glass bottle 5g Each for $81.29
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 165.19
Quantity 5g
Color Clear yellow
EINECS Number 220-365-1
Density 1.084g/mL
Chemical Name or Material Ethyl 2-pyridylacetate
Refractive Index 1.4963 to 1.4983 (20°C, 589nm)
Name Note 98%
Linear Formula (C5H4N)CH2COOC2H5
MDL Number MFCD00006359
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H11NO2
CAS 2739-98-2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.