missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Piperidinecarboxylate, TCI America™

Manufacturer:  TCI America E08375G

 View more versions of this product

Catalog No. E08375G



Specifications

Specifications

Ethyl 2-Piperidinecarboxylate
Yellow
15862-72-3
MFCD00005980
ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
CCOC(=O)C1CCCCN1
157.213
157.21
≥98.0% (GC,T)
217°C
5g
C8H15NO2
1993
SZIKRGHFZTYTIT-UHFFFAOYSA-N
ethyl piperidine-2-carboxylate
27517
Clear Liquid at 20°C

Chemical Identifiers

15862-72-3
157.213
SZIKRGHFZTYTIT-UHFFFAOYSA-N
27517
CCOC(=O)C1CCCCN1
C8H15NO2
MFCD00005980
ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
ethyl piperidine-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents