Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-phenylacetoacetate, 97%

$65.09 - $213.96

Chemical Identifiers

Chemical Name or Material Ethyl 2-phenylacetoacetate
CAS 5413-05-8
Molecular Formula C12H14O3
Formula Weight 206.24
MDL Number MFCD00040490
Beilstein 1912655
Synonym 2-Phenylacetoacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1625703

  SDS
alfa aesar™
L1625703
1g Each for $65.09

AAL1625706

  SDS
alfa aesar™
L1625706
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 206.24
Chemical Name or Material Ethyl 2-phenylacetoacetate
MDL Number MFCD00040490
CAS 5413-05-8
Assay 97%
Molecular Formula C12H14O3
Beilstein 1912655
Synonym 2-Phenylacetoacetic acid ethyl ester
Odor Odorless
Boiling Point 140° to 144°C (10mmHg)
Quantity 1g