Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-pentynyl ether, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-pentynyl ether
CAS 20635-10-3
Molecular Formula C7H12O
Linear Formula CH3CH2C≡CCH2OCH2CH3
Formula Weight 112.17
MDL Number MFCD00048702
UN Number UN3271
Synonym 1-Ethoxy-2-pentyne
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2596006

  SDS
alfa aesar™
H2596006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2596014

  SDS
alfa aesar™
H2596014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.17
Chemical Name or Material Ethyl 2-pentynyl ether
Assay 97%
Molecular Formula C7H12O
Linear Formula CH3CH2C≡CCH2OCH2CH3
MDL Number MFCD00048702
CAS 20635-10-3
UN Number UN3271
Synonym 1-Ethoxy-2-pentyne
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Flash Point 30°C (86°F)
Boiling Point 63° to 65°C (20mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.