Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-oxopyrrolidine-4-carboxylate, 96%

Chemical Identifiers

Chemical Name or Material Ethyl 2-oxopyrrolidine-4-carboxylate
CAS 60298-18-2
Formula Weight 157.17
MDL Number MFCD08274999
Synonym 2-Oxopyrrolidine-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6615703

 
alfa aesar™
H6615703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66157MD

 
alfa aesar™
H66157MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.17
Melting Point 40° to 47°C
Chemical Name or Material Ethyl 2-oxopyrrolidine-4-carboxylate
MDL Number MFCD08274999
Physical Form Solid
Assay 96%
CAS 60298-18-2
Synonym 2-Oxopyrrolidine-4-carboxylic acid ethyl ester
Color Pale brown to dark brown
Quantity 1g