Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-oxocyclopentylacetate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 2-oxocyclopentylacetate
CAS 20826-94-2
Formula Weight 170.21
MDL Number MFCD00044715
Synonym 2-Oxocyclopentylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6158906

  SDS
alfa aesar™
H6158906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6158914

  SDS
alfa aesar™
H6158914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.21
CAS 20826-94-2
Assay 95%
Quantity 5g
MDL Number MFCD00044715
Physical Form Solid
Refractive Index 1.4517
Chemical Name or Material Ethyl 2-oxocyclopentylacetate
Storage Note 1 -30° to -10°C
Synonym 2-Oxocyclopentylacetic acid ethyl ester

Applications
Ethyl 2-oxocyclopentylacetate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Safety and Handling

-30° to -10°C