missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-oxocyclopentylacetate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6158906
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEthyl 2-oxocyclopentylacetate is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Ethyl 2-oxocyclopentylacetate | |
C9H14O3 | |
MFCD00044715 | |
ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate | |
PJMKFKUFBDXYEC-SSDOTTSWSA-N | |
ethyl 2-(2-oxocyclopentyl)acetate | |
2723663 | |
95% |
20826-94-2 | |
1.4517 | |
5 g | |
Slightly soluble in water. | |
CCOC(=O)C[C@H]1CCCC1=O | |
170.21 | |
170.21 | |
Solid |
Chemical Identifiers
20826-94-2 | |
170.21 | |
PJMKFKUFBDXYEC-SSDOTTSWSA-N | |
2723663 | |
CCOC(=O)C[C@H]1CCCC1=O |
C9H14O3 | |
MFCD00044715 | |
ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate | |
ethyl 2-(2-oxocyclopentyl)acetate |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C