missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-oxo-4-phenylbutyrate, 90+%, Thermo Scientific Chemicals
$85.45 - $269.26
Chemical Identifiers
CAS | 64920-29-2 |
---|---|
Molecular Formula | C12H14O3 |
Molecular Weight (g/mol) | 206.241 |
MDL Number | MFCD00037533 |
InChI Key | STPXIOGYOLJXMZ-UHFFFAOYSA-N |
Synonym | ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate |
PubChem CID | 562087 |
IUPAC Name | ethyl 2-oxo-4-phenylbutanoate |
SMILES | CCOC(=O)C(=O)CCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2014809
|
Thermo Scientific Chemicals
B2014809 |
10 g |
N/A
|
|
|||||
AAB2014818
|
Thermo Scientific Chemicals
B2014818 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64920-29-2 | |
206.241 | |
STPXIOGYOLJXMZ-UHFFFAOYSA-N | |
562087 | |
CCOC(=O)C(=O)CCC1=CC=CC=C1 |
C12H14O3 | |
MFCD00037533 | |
ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate | |
ethyl 2-oxo-4-phenylbutanoate |
Specifications
64920-29-2 | |
131°C to 133°C (2 mmHg) | |
C12H14O3 | |
MFCD00037533 | |
2725083 | |
STPXIOGYOLJXMZ-UHFFFAOYSA-N | |
ethyl 2-oxo-4-phenylbutanoate | |
562087 | |
≥90% |
1.09 | |
>110°C (230°F) | |
1.502 | |
10 g | |
ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate | |
CCOC(=O)C(=O)CCC1=CC=CC=C1 | |
206.241 | |
206.24 | |
Ethyl 2-oxo-4-phenylbutyrate |
Safety and Handling
EINECSNumber : 265-276-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only