Please login to your online account to display your discounted pricing

Ethyl 2-oximinooxamate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426880010

 View more versions of this product

Catalog No. AC426880010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 132.12
Quantity 1g
Color White to yellow
Melting Point 96° to 103°C
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C4H8N2O3
Chemical Name or Material Ethyl 2-oximinooxamate
CAS 10489-74-4
Linear Formula HONHC(=NH)COOCH2CH3
MDL Number MFCD03013442
Infrared Spectrum Authentic
Health Hazard 1 Warning

Specifications

Chemical Name or Material Ethyl 2-oximinooxamate
Assay ≥96.0%
Assay Percent Range 98%
Molecular Formula C4H8N2O3
Linear Formula HONHC(=NH)COOCH2CH3
Formula Weight 132.12
MDL Number MFCD03013442
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

SDS & Safety Documents