Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-octynoate, 98%

$49.11 - $574.08

Chemical Identifiers

Chemical Name or Material Ethyl 2-octynoate
CAS 10519-20-7
Molecular Formula C10H16O2
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
Formula Weight 168.24
MDL Number MFCD00015286
Beilstein 1759822
Synonym 2-Octynoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1978406

  SDS
alfa aesar™
A1978406
5g Each for $49.11

AAA1978422

  SDS
alfa aesar™
A1978422
100g Each for $574.08
Description & Specifications

Specifications

Formula Weight 168.24
Boiling Point 85°C (6mmHg)
Synonym 2-Octynoic acid ethyl ester
Molecular Formula C10H16O2
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
MDL Number MFCD00015286
Chemical Name or Material Ethyl 2-octynoate
Beilstein 1759822
Recommended Storage 2° to 8°C
TSCA TSCA
Assay 98%
Quantity 5g
Flash Point 99°C (210°F)
CAS 10519-20-7
Safety and Handling

2° to 8°C