Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(N-phthalimidooxy)-2-methylpropionate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 2-(N-phthalimidooxy)-2-methylpropionate
CAS 40674-21-3
Formula Weight 277.27
Synonym Ethyl 2-(1,3-dioxo-1,3-dihydro-2-isoindolyloxy)-2-methylpropionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6166003

 
alfa aesar™
H6166003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61660MD

 
alfa aesar™
H61660MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.27
Chemical Name or Material Ethyl 2-(N-phthalimidooxy)-2-methylpropionate
Physical Form Solid
Assay 98%
CAS 40674-21-3
Synonym Ethyl 2-(1,3-dioxo-1,3-dihydro-2-isoindolyloxy)-2-methylpropionate
Quantity 1g