Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-n-butylacetoacetate, 95%

$72.80 - $72.80

Chemical Identifiers

Chemical Name or Material Ethyl 2-n-butylacetoacetate
CAS 1540-29-0
Molecular Formula C10H18O3
Formula Weight 186.25
MDL Number MFCD00026888
Beilstein 636718
Synonym Ethyl 2-acetylhexanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0090506

 
alfa aesar™
L0090506
5g Each for $72.80
Description & Specifications

Specifications

Formula Weight 186.25
Chemical Name or Material Ethyl 2-n-butylacetoacetate
Assay 95%
MDL Number MFCD00026888
CAS 1540-29-0
Molecular Formula C10H18O3
Beilstein 636718
Synonym Ethyl 2-acetylhexanoate
Boiling Point 224°C
Quantity 5g