Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-methylthiazole-4-carboxylate, 98%

$302.86 - $302.86

Chemical Identifiers

Chemical Name or Material Ethyl 2-methylthiazole-4-carboxylate
CAS 6436-59-5
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00156148
Synonym 2-Methylthiazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1205006

  SDS
alfa aesar™
A1205006
5g Each for $302.86
Description & Specifications

Specifications

Formula Weight 171.22
Assay 98%
CAS 6436-59-5
Synonym 2-Methylthiazole-4-carboxylic acid ethyl ester
Health Hazard 1 Warning
Chemical Name or Material Ethyl 2-methylthiazole-4-carboxylate
Melting Point 54° to 56°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00156148
Molecular Formula C7H9NO2S
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.