Please login to your online account to display your discounted pricing

Ethyl 2-methylphenylacetate, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-methylphenylacetate
Name Note 96%
CAS 40291-39-2
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C11H14O2
Linear Formula CH3C6H4CH2CO2C2H5
Formula Weight 178.23
MDL Number MFCD00009179
Synonym Ethyl 2-methylbenzeneacetate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC279310250

  SDS
thermo fisher scientific
279310250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 178.23
Physical Form Liquid
Quantity 25g
Color Clear colorless
Beilstein 09, 2426
EINECS Number 254-871-9
Density 0.9980g/mL
Refractive Index 1.5005 to 1.5025 (20°C, 589nm)
Synonym Ethyl 2-methylbenzeneacetate
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Assay Percent Range 96%
Linear Formula CH3C6H4CH2CO2C2H5
Chemical Name or Material Ethyl 2-methylphenylacetate
Name Note 96%
Molecular Formula C11H14O2
MDL Number MFCD00009179
CAS 40291-39-2
Safety and Handling

GHS P Statement:
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.