Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (2-methylphenoxy)acetate, 99%

$90.72 - $907.07

Chemical Identifiers

Chemical Name or Material Ethyl (2-methylphenoxy)acetate
CAS 93917-68-1
Molecular Formula C11H14O
Formula Weight 194.23
MDL Number MFCD00216559
Synonym (2-Methylphenoxy)acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2237506

  SDS
alfa aesar™
B2237506
5g Each for $90.72

AAB2237514

  SDS
alfa aesar™
B2237514
25g Each for $311.91

AAB2237522

  SDS
alfa aesar™
B2237522
100g Each for $907.07
Description & Specifications

Specifications

Formula Weight 194.23
Synonym (2-Methylphenoxy)acetic acid ethyl ester
MDL Number MFCD00216559
Flash Point >110°C (230°F)
Chemical Name or Material Ethyl (2-methylphenoxy)acetate
Boiling Point 119° to 120°C
CAS 93917-68-1
Quantity 5g
Molecular Formula C11H14O
Assay 99%