missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Methylnicotinate, TCI America™

Manufacturer:  TCI America E086025G

 View more versions of this product

Catalog No. E086025G



Specifications

Specifications

Ethyl 2-Methylnicotinate
Yellow
1721-26-2
MFCD00006336
TZORNSWZUZDUCU-UHFFFAOYSA-N
ethyl 2-methylpyridine-3-carboxylate
74401
Clear Liquid at 20°C
125°C
25g
C9H11NO2
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727
CCOC(=O)C1=C(N=CC=C1)C
165.192
165.19
≥98.0% (GC,T)

Chemical Identifiers

1721-26-2
165.192
TZORNSWZUZDUCU-UHFFFAOYSA-N
74401
CCOC(=O)C1=C(N=CC=C1)C
C9H11NO2
MFCD00006336
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727
ethyl 2-methylpyridine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents