missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Methylacetoacetate, TCI America™

Manufacturer:  TCI America M069825ML

 View more versions of this product

Catalog No. M069825ML



Specifications

Specifications

Ethyl 2-Methylacetoacetate
Yellow
609-14-3
MFCD00009164
FNENWZWNOPCZGK-UHFFFAOYSA-N
ethyl 2-methyl-3-oxobutanoate
701
Clear Liquid at 20°C
185°C
25mL
C7H12O3
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
CCOC(=O)C(C)C(=O)C
144.17
144.17
≥95.0% (GC)

Chemical Identifiers

609-14-3
144.17
FNENWZWNOPCZGK-UHFFFAOYSA-N
701
CCOC(=O)C(C)C(=O)C
C7H12O3
MFCD00009164
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
ethyl 2-methyl-3-oxobutanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1505

TSCA : Yes

Documents