Learn More
Ethyl 2-methylacetoacetate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1162914
Description
Ethyl 2-methylacetoacetate is used in Japp-Klingemann reaction to prepare hydrazones from beta-keto acid and aryl diazonium salts. It is used as a precursor involved in the synthesis of ethylpyruvate phenylhydrazone via reaction with benzenediazonium chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-methylacetoacetate is used in Japp-Klingemann reaction to prepare hydrazones from beta-keto acid and aryl diazonium salts. It is used as a precursor involved in the synthesis of ethylpyruvate phenylhydrazone via reaction with benzenediazonium chloride.
Solubility
Miscible with alcohol and ether. Silghtly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Ethyl 2-methylacetoacetate | |
-45°C | |
185°C | |
Ester-like | |
1.418 | |
25 g | |
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester | |
FNENWZWNOPCZGK-UHFFFAOYSA-N | |
ethyl 2-methyl-3-oxobutanoate | |
701 | |
95% |
609-14-3 | |
1.019 | |
62°C (143°F) | |
C7H12O3 | |
MFCD00009164 | |
1071742 | |
Miscible with alcohol and ether. Silghtly miscible with water. | |
CCOC(=O)C(C)C(=O)C | |
144.17 | |
144.17 |
Chemical Identifiers
609-14-3 | |
144.17 | |
FNENWZWNOPCZGK-UHFFFAOYSA-N | |
701 | |
CCOC(=O)C(C)C(=O)C |
C7H12O3 | |
MFCD00009164 | |
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester | |
ethyl 2-methyl-3-oxobutanoate |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 210-179-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only