Learn More
Ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC42223EA
Specifications
Ethyl 2-methyl-4-phenyl-1, 3-thiazole-5-carboxylate | |
96.5 | |
97% | |
MFCD07346320 | |
ethyl 2-methyl-4-phenylthiazole-5-carboxylate, 2-methyl-4-phenylthiazole-5-carboxylic acid ethyl ester, 2-methyl-4-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl-, ethyl ester | |
CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1 | |
247.31 | |
247.32 |
32043-95-1 | |
100.0 | |
C13H13NO2S | |
10g | |
UHLMXNFHHFDVPW-UHFFFAOYSA-N | |
ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate | |
7131013 | |
97% |
Chemical Identifiers
32043-95-1 | |
247.31 | |
UHLMXNFHHFDVPW-UHFFFAOYSA-N | |
7131013 | |
CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1 |
C13H13NO2S | |
MFCD07346320 | |
ethyl 2-methyl-4-phenylthiazole-5-carboxylate, 2-methyl-4-phenylthiazole-5-carboxylic acid ethyl ester, 2-methyl-4-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl-, ethyl ester | |
ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning