Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-methyl-2-(methylthio)propionate, 98%

$186.84 - $186.84

Chemical Identifiers

Chemical Name or Material Ethyl 2-methyl-2-(methylthio)propionate
CAS 49773-24-2
Molecular Formula C7H14O2S
Formula Weight 162.25
MDL Number MFCD00053099
Synonym Ethyl 2-(methylthio)isobutyrate; 2-Methyl-2-(methylthio)propionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2259709

  SDS
alfa aesar™
B2259709
10g Each for $186.84
Description & Specifications

Specifications

Formula Weight 162.25
Chemical Name or Material Ethyl 2-methyl-2-(methylthio)propionate
Assay 98%
Molecular Formula C7H14O2S
MDL Number MFCD00053099
CAS 49773-24-2
Synonym Ethyl 2-(methylthio)isobutyrate; 2-Methyl-2-(methylthio)propionic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.