Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate, 97%

$48.49 - $48.49

Chemical Identifiers

Chemical Name or Material Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate
CAS 3652-61-7
Molecular Formula C20H19NO2
Formula Weight 305.38
MDL Number MFCD00221065
Synonym 2-Methyl-1,5-diphenylpyrrole-3-carboxylic acid ethyl ester; 1,5-Diphenyl-2-methylpyrrole-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1983203

  SDS
alfa aesar™
A1983203
1g Each for $48.49
Description & Specifications

Specifications

Formula Weight 305.38
Chemical Name or Material Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate
Assay 97%
Molecular Formula C20H19NO2
MDL Number MFCD00221065
CAS 3652-61-7
Synonym 2-Methyl-1,5-diphenylpyrrole-3-carboxylic acid ethyl ester; 1,5-Diphenyl-2-methylpyrrole-3-carboxylic acid ethyl ester
Melting Point 95° to 99°C
Odor Odorless
Quantity 1g