missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (2-Methoxyphenoxy)acetate, TCI America™

Manufacturer:  TCI America E124125G

 View more versions of this product

Catalog No. E124125G



Specifications

Specifications

Ethyl (2-Methoxyphenoxy)acetate
Yellow
13078-21-2
MFCD00082657
BZCGGCRVJFWCIW-UHFFFAOYSA-N
ethyl 2-(2-methoxyphenoxy)acetate
83114
Clear Liquid at 20°C
116°C
25g
C11H14O4
(2-Methoxyphenoxy)acetic Acid Ethyl Ester
CCOC(=O)COC1=CC=CC=C1OC
210.229
210.23
≥98.0% (GC)

Chemical Identifiers

13078-21-2
210.229
BZCGGCRVJFWCIW-UHFFFAOYSA-N
83114
CCOC(=O)COC1=CC=CC=C1OC
C11H14O4
MFCD00082657
(2-Methoxyphenoxy)acetic Acid Ethyl Ester
ethyl 2-(2-methoxyphenoxy)acetate
Safety and Handling

Safety and Handling

TSCA : No

Documents