Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-methoxybenzoate, 98+%

$34.49 - $520.27

Chemical Identifiers

Chemical Name or Material Ethyl 2-methoxybenzoate
CAS 7335-26-4
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00015148
Beilstein 2575515
Synonym Ethyl o-anisate; 2-Methoxybenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1128718

  SDS
alfa aesar™
A1128718
50g Each for $124.68

AAA1128709

  SDS
alfa aesar™
A1128709
10g Each for $34.49

AAA1128730

  SDS
alfa aesar™
A1128730
250g Each for $520.27
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material Ethyl 2-methoxybenzoate
TSCA TSCA
Assay ≥98%
Molecular Formula C10H12O3
MDL Number MFCD00015148
CAS 7335-26-4
Beilstein 2575515
Synonym Ethyl o-anisate; 2-Methoxybenzoic acid ethyl ester
Boiling Point 127° to 128°C (11mmHg)
Quantity 50g