Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-mercaptopropionate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 2-mercaptopropionate
CAS 19788-49-9
Molecular Formula C5H10O2S
Formula Weight 134.2
MDL Number MFCD00040233
UN Number UN1993
Beilstein 1071465
Synonym Ethyl thiolactate; 2-Mercaptopropionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1152006

  SDS
alfa aesar™
L1152006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1152014

  SDS
alfa aesar™
L1152014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.2
CAS 19788-49-9
Molecular Formula C5H10O2S
Assay 98%
Health Hazard 1 Danger
MDL Number MFCD00040233
Health Hazard 2 GHS H Statement
H301-H311-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1993
Chemical Name or Material Ethyl 2-mercaptopropionate
Quantity 5g
Flash Point 46°C (115°F)
Boiling Point 75° to 76°C (35mmHg)
Synonym Ethyl thiolactate; 2-Mercaptopropionic acid ethyl ester
Odor Strong
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Beilstein 1071465
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.