missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Isopropylacetoacetate, TCI America™

Manufacturer:  TCI America E11195ML

 View more versions of this product

Catalog No. E11195ML



Specifications

Specifications

Ethyl 2-Isopropylacetoacetate
Colorless
1522-46-9
MFCD00042884
DMIFFKCVURTPTG-UHFFFAOYSA-N
ethyl 2-acetyl-3-methylbutanoate
95544
Clear Liquid at 20°C
202°C
5mL
C9H16O3
ethyl 2-isopropylacetoacetate, 2-isopropylacetoacetic acid ethyl ester, ethyl 2-iso-propylacetoacetate, butanoic acid, 2-acetyl-3-methyl-, ethyl ester, ethyl alpha-isopropylacetoacetate, ethyl 2-acetyl-3-methylbutyrate, acetoacetic acid, 2-isopropyl-, ethyl ester, ethyl .alpha.-isopropylacetoacetate, ethyl isopropylaceto-acetate, acmc-1c35s
CCOC(=O)C(C(C)C)C(=O)C
172.224
172.22
≥94.0% (GC)

Chemical Identifiers

1522-46-9
172.224
DMIFFKCVURTPTG-UHFFFAOYSA-N
95544
CCOC(=O)C(C(C)C)C(=O)C
C9H16O3
MFCD00042884
ethyl 2-isopropylacetoacetate, 2-isopropylacetoacetic acid ethyl ester, ethyl 2-iso-propylacetoacetate, butanoic acid, 2-acetyl-3-methyl-, ethyl ester, ethyl alpha-isopropylacetoacetate, ethyl 2-acetyl-3-methylbutyrate, acetoacetic acid, 2-isopropyl-, ethyl ester, ethyl .alpha.-isopropylacetoacetate, ethyl isopropylaceto-acetate, acmc-1c35s
ethyl 2-acetyl-3-methylbutanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1505

TSCA : Yes

Documents