Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-isocyanato-4-methylvalerate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-isocyanato-4-methylvalerate
CAS 64505-10-8
Molecular Formula C9H15NO3
Formula Weight 185.22
MDL Number MFCD00078351
UN Number UN2206
Beilstein 1240785
Synonym Ethyl 2-isocyanato-4-methylpentanoate; 2-Isocyanato-4-methylvaleric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1161303

 
alfa aesar™
L1161303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1161306

 
alfa aesar™
L1161306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.22
Chemical Name or Material Ethyl 2-isocyanato-4-methylvalerate
Assay 97%
Molecular Formula C9H15NO3
MDL Number MFCD00078351
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 64505-10-8
UN Number UN2206
Beilstein 1240785
Synonym Ethyl 2-isocyanato-4-methylpentanoate; 2-Isocyanato-4-methylvaleric acid ethyl ester
Boiling Point 92° to 93°C (8mmHg)
Quantity 1g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C