Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-isocyanato-4-(methylthio)butyrate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-isocyanato-4-(methylthio)butyrate
CAS 64505-12-0
Molecular Formula C8H13NO3S
Formula Weight 203.26
MDL Number MFCD00078433
UN Number UN2206
Beilstein 1710840
Synonym 2-Isocyanato-4-(methylthio)butyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1305904

 
alfa aesar™
L1305904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1305909

 
alfa aesar™
L1305909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.26
MDL Number MFCD00078433
CAS 64505-12-0
Synonym 2-Isocyanato-4-(methylthio)butyric acid ethyl ester
Chemical Name or Material Ethyl 2-isocyanato-4-(methylthio)butyrate
Assay 97%
Molecular Formula C8H13NO3S
UN Number UN2206
Beilstein 1710840
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 155°C (24mmHg)
Quantity 2g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C