Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-isocyanato-3-methylbutyrate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-isocyanato-3-methylbutyrate
CAS 5296-78-6
Molecular Formula C8H13NO3
Formula Weight 171.2
MDL Number MFCD00078478
UN Number UN2206
Beilstein 1239965
Synonym 2-Isocyanato-3-methylbutyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0998809

  SDS
alfa aesar™
L0998809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0998804

  SDS
alfa aesar™
L0998804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.2
Chemical Name or Material Ethyl 2-isocyanato-3-methylbutyrate
Assay 97%
Molecular Formula C8H13NO3
MDL Number MFCD00078478
CAS 5296-78-6
Beilstein 1239965
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 81° to 82°C (3mmHg)
UN Number UN2206
Synonym 2-Isocyanato-3-methylbutyric acid ethyl ester
Odor Irritating
Quantity 10g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C