missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-Iodopropionate 97.0+%, TCI America™
Supplier: TCI America E119825G
Specifications
Ethyl 2-Iodopropionate | |
79°C | |
MFCD02258628 | |
2-Iodopropionic Acid Ethyl Ester | |
CCOC(=O)C(C)I | |
228.029 | |
228.03 | |
Liquid |
31253-08-4 | |
C5H9IO2 | |
25 g | |
AVMMXNKUHBWIMU-UHFFFAOYSA-N | |
ethyl 2-iodopropanoate | |
10922254 | |
≥97.0% (GC) |
Chemical Identifiers
31253-08-4 | |
228.029 | |
AVMMXNKUHBWIMU-UHFFFAOYSA-N | |
10922254 | |
CCOC(=O)C(C)I |
C5H9IO2 | |
MFCD02258628 | |
2-Iodopropionic Acid Ethyl Ester | |
ethyl 2-iodopropanoate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer