missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Iodo-2-methylpropionate, TCI America™

Manufacturer:  TCI America E119925G

 View more versions of this product

Catalog No. E119925G



Specifications

Specifications

Ethyl 2-Iodo-2-methylpropionate
25g
C6H11IO2
AYVTVRNTNCEPSH-UHFFFAOYSA-N
ethyl 2-iodo-2-methylpropanoate
11299482
Clear Liquid at 20°C
Undesignated
7425-55-0
2-Iodo-2-methylpropionic Acid Ethyl Ester
CCOC(=O)C(C)(C)I
242.056
242.06
≥94.0% (GC)

Chemical Identifiers

7425-55-0
242.056
2-Iodo-2-methylpropionic Acid Ethyl Ester
ethyl 2-iodo-2-methylpropanoate
C6H11IO2
AYVTVRNTNCEPSH-UHFFFAOYSA-N
11299482
CCOC(=O)C(C)(C)I
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents