missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-hydroxyisobutyrate, 98%

Manufacturer:  Alfa Aesar B2359222

 View more versions of this product

Catalog No. AAB2359222


 

Specifications

Specifications

Ethyl 2-hydroxyisobutyrate
98%
MFCD00004458
878308
GFUIDHWFLMPAGY-UHFFFAOYSA-N
ethyl 2-hydroxy-2-methylpropanoate
6653
149°C to 150°C
44°C (111°F)
80-55-7
C6H12O3
UN3272
Ethyl 2-hydroxy-2-methylpropionate, Ethyl 2-methyllactate
CCOC(=O)C(C)(C)O
132.159
132.16
100g

Chemical Identifiers

80-55-7
132.159
GFUIDHWFLMPAGY-UHFFFAOYSA-N
6653
C6H12O3
MFCD00004458
Ethyl 2-hydroxy-2-methylpropionate, Ethyl 2-methyllactate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : OD5672000

SDS Documents