Learn More
Thermo Scientific Chemicals Ethyl 2-hydroxyethyl sulfide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 118551000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 2-hydroxyethyl sulfide | |
110-77-0 | |
Colorless to Yellow | |
180°C to 184°C | |
98% | |
C4H10OS | |
MFCD00002909 | |
Glass bottle | |
1.4859 to 1.4879 | |
2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol | |
LNRIEBFNWGMXKP-UHFFFAOYSA-N | |
2-ethylsulfanylethanol | |
8075 | |
Liquid |
98% | |
1.0200g/mL | |
-100°C | |
93°C | |
Authentic | |
C2H5SCH2CH2OH | |
01, II, 525 | |
15, 3906 | |
100g | |
1.02 | |
CCSCCO | |
106.18 | |
106.18 | |
98% |
Chemical Identifiers
110-77-0 | |
106.18 | |
LNRIEBFNWGMXKP-UHFFFAOYSA-N | |
8075 | |
CCSCCO |
C4H10OS | |
MFCD00002909 | |
2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol | |
2-ethylsulfanylethanol |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 203-802-0
RTECSNumber : KL1225000
TSCA : TSCA
RUO â Research Use Only