Learn More
Ethyl 2-hydroxyethyl sulfide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118550050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-hydroxyethyl sulfide | |
110-77-0 | |
180°C to 184°C | |
Authentic | |
Glass bottle | |
1.4859 to 1.4879 | |
MFCD00002909 | |
01, II, 525 | |
15, 3906 | |
LNRIEBFNWGMXKP-UHFFFAOYSA-N | |
2-ethylsulfanylethanol | |
8075 | |
98% |
98% | |
1.0200g/mL | |
93°C | |
98% | |
C4H10OS | |
C2H5SCH2CH2OH | |
5g | |
1.02 | |
2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol | |
CCSCCO | |
106.18 | |
106.18 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 203-802-0
RTECSNumber : KL1225000
TSCA : TSCA
RUO â Research Use Only