Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-hexynoate, 98+%

$54.03 - $54.03

Chemical Identifiers

Chemical Name or Material Ethyl 2-hexynoate
CAS 16205-90-6
Molecular Formula C8H12O2
Linear Formula CH3CH2CH2C≡CCO2CH2CH3
Formula Weight 140.18
MDL Number MFCD00015257
Beilstein 1702264
Synonym 2-Hexynoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0523704

 
alfa aesar™
L0523704
2g Each for $54.03
Description & Specifications

Specifications

Formula Weight 140.18
Chemical Name or Material Ethyl 2-hexynoate
CAS 16205-90-6
Synonym 2-Hexynoic acid ethyl ester
Assay ≥98%
Molecular Formula C8H12O2
Linear Formula CH3CH2CH2C≡CCO2CH2CH3
MDL Number MFCD00015257
Beilstein 1702264
Boiling Point 87° to 88°C (14mmHg)
Quantity 2g
Recommended Storage 2° to 8°C

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Safety and Handling

2° to 8°C